2-Methyl-5-(pyrrolidin-2-yl)pyridine 97%

Lieferant: Apollo Scientific

OR350511-250MG
APOSOR350511-250MGEA 242 EUR
APOSOR350511-250MG
2-Methyl-5-(pyrrolidin-2-yl)pyridine 97%
2-Methyl-5-(pyrrolidin-2-yl)pyridine
Formel: C₁₀H₁₄N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12024458
CAS-Nummer: 90872-72-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR