3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-tosyl-1H-pyrrolo[2,3-b]pyridine 97%

Lieferant: Apollo Scientific

OR350552-250MG
APOSOR350552-250MGEA 105 EUR
APOSOR350552-250MG
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-tosyl-1H-pyrrolo[2,3-b]pyridine 97%
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-tosyl-1H-pyrrolo[2,3-b]pyridine
Formel: C₂₀H₂₃BN₂O₄S
Molecular Weight: 398,284 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11616456
CAS-Nummer: 866545-91-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR