4-(2-Hydroxy-3-morpholin-4-ylpropoxy)benzeneboronic acid, pinacol ester 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(Morpholin-1-yl)-3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy]propan-2-ol

OR3510-100MG
APOSOR3510-100MGEA 87 EUR
APOSOR3510-100MG
4-(2-Hydroxy-3-morpholin-4-ylpropoxy)benzeneboronic acid, pinacol ester 97%
4-(2-Hydroxy-3-morpholin-4-ylpropoxy)benzeneboronic acid, pinacol ester
Formel: C₁₉H₃₀BNO₅
Molecular Weight: 363,256 g/mol
Schmelzpunkt: 121…125 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09258653
CAS-Nummer: 756520-73-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR