3-Cyan-7-ethoxycumarin 98+%

Lieferant: Apollo Scientific

OR351016-250MG
APOSOR351016-250MGEA 571 EUR
APOSOR351016-250MG
3-Cyan-7-ethoxycumarin 98+%
3-Cyan-7-ethoxycumarin
Formel: C₁₂H₉NO₃
Molecular Weight: 215,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00467359
CAS-Nummer: 117620-77-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR