7-Diethylamino-3-formylcoumarin

Lieferant: Apollo Scientific

Synonyme: 7-(Diethylamino)-2-oxo-2H-chromene-3-carbaldehyde

OR351018-250MG
APOSOR351018-250MGEA 116 EUR
APOSOR351018-250MG
7-Diethylamino-3-formylcoumarin
7-Diethylamino-3-formylcoumarin
Formel: C₁₄H₁₅NO₃
Molecular Weight: 245,274 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00438856
CAS-Nummer: 57597-64-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR