2-Isopropyl-N,N,6-trimethylanilin 95%

Lieferant: Apollo Scientific
Achtung

OR3518-1G
APOSOR3518-1GEA 109 EUR
APOSOR3518-1G
2-Isopropyl-N,N,6-trimethylanilin 95%
2-Isopropyl-N,N,6-trimethylanilin
Formel: C₁₂H₁₉N
Siedepunkt: 102 °C/12mmHg
Lagertemperatur: Raumtemperatur
CAS-Nummer: 227199-09-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR