Methyl-(3-bromphenyl)acetat 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Bromophenylacetic acid methyll ester, Methyl 3-Bromophenylacetate, Methyl 2-(3-bromophenyl)acetate
OR3537-100G
OR3537-25G
OR3537-5G
APOSOR3537-100GEA
248
EUR
APOSOR3537-100G
APOSOR3537-25G
APOSOR3537-5G
Methyl-(3-bromphenyl)acetat 98%
Methyl-(3-bromphenyl)acetat
Formel:
C₉H₉BrO₂ Molecular Weight: 229.0700 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06858764 CAS-Nummer: 150529-73-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...