Isochinolin-8-amin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Isoquinolin-8-amine, 8-Aminoisoquinoline
OR3549-1G
APOSOR3549-1GEA
31.9
EUR
APOSOR3549-1G
Isochinolin-8-amin 98%
Isochinolin-8-amin
Formel:
C₉H₈N₂ Molecular Weight: 144.1700 g/mol Schmelzpunkt: 168…169 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00179553 CAS-Nummer: 23687-27-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...