Methyl-3-(chlorsulfonyl)benzoat
Lieferant: Apollo Scientific
|
Gefahr
|
OR3564-1G
OR3564-25G
OR3564-5G
APOSOR3564-1GEA
0
EUR
APOSOR3564-1G
APOSOR3564-25G
APOSOR3564-5G
Methyl-3-(chlorsulfonyl)benzoat
Methyl-3-(chlorsulfonyl)benzoat
Formel:
C₈H₇ClO₄S Schmelzpunkt: 63…69 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06408800 CAS-Nummer: 63555-50-0 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...