2-Methoxy-5-(methylsulphonyl)benzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Carboxy-4-(methylsulphonyl)anisole, 5-(Methylsulphonyl)-o-anisic acid, 3-Carboxy-4-methoxyphenyl methyl sulphone

OR3569-25G OR3569-5G OR3569-100G
APOSOR3569-25GEA 48 EUR
APOSOR3569-25G APOSOR3569-5G APOSOR3569-100G
2-Methoxy-5-(methylsulphonyl)benzoic acid
2-Methoxy-5-(methylsulphonyl)benzoic acid
Formel: C₉H₁₀O₅S
Schmelzpunkt: 190…193 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01317544
CAS-Nummer: 50390-76-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR