2,5-Dibrom-3-nitropyridin 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR3579-100G
OR3579-25G
APOSOR3579-100GEA
0
EUR
APOSOR3579-100G
APOSOR3579-25G
2,5-Dibrom-3-nitropyridin 98%
2,5-Dibromo-3-nitropyridine
Formel:
C₅H₂Br₂N₂O₂ Schmelzpunkt: 94 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09266223 CAS-Nummer: 15862-37-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...