5-Brom-2-chlor-3-nitropyridin

Lieferant: Apollo Scientific
Achtung

OR3580-25G OR3580-100G OR3580-5G
APOSOR3580-25GEA 26.4 EUR
APOSOR3580-25G APOSOR3580-100G APOSOR3580-5G
5-Brom-2-chlor-3-nitropyridin
5-Brom-2-chlor-3-nitropyridin
Formel: C₅H₂BrClN₂O₂
Molecular Weight: 237.44 g/mol
Schmelzpunkt: 65…70 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00222270
CAS-Nummer: 67443-38-3
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR