5-Brom-2-chlor-3-nitropyridin
Lieferant: Apollo Scientific
|
Achtung
|
OR3580-5G
APOSOR3580-5GEA
21.9
EUR
APOSOR3580-5G
5-Brom-2-chlor-3-nitropyridin
5-Brom-2-chlor-3-nitropyridin
Formel:
C₅H₂BrClN₂O₂ Molecular Weight: 237,44 g/mol Schmelzpunkt: 65…70 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00222270 CAS-Nummer: 67443-38-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...