3-Brom-2,5-dichlorpyridin

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-2,5-dichloro-pyridine

OR3581-5G OR3581-1G OR3581-25G
APOSOR3581-5GEA 26.1 EUR
APOSOR3581-5G APOSOR3581-1G APOSOR3581-25G
3-Brom-2,5-dichlorpyridin
3-Bromo-2,5-dichloropyridine
Formel: C₅H₂BrCl₂N
Molecular Weight: 226.89 g/mol
Schmelzpunkt: 38…40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659484
CAS-Nummer: 138006-41-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR