5-Oxazolcarbonsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Oxazole-5-carboxylic acid

OR3589-500MG OR3589-25G OR3589-5G
APOSOR3589-500MGEA 36.4 EUR
APOSOR3589-500MG APOSOR3589-25G APOSOR3589-5G
5-Oxazolcarbonsäure 98%
5-Oxazolecarboxylic acid
Formel: C₄H₃NO₃
Molecular Weight: 113.07 g/mol
Schmelzpunkt: 226…230 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD04114931
CAS-Nummer: 118994-90-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR