3-[(4-Methoxybenzyl)sulphamoyl]benzeneboronic acid 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-4-Methoxybenzyl-3-borono benzenesulphonamide

OR3592-250MG
APOSOR3592-250MGEA 109 EUR
APOSOR3592-250MG
3-[(4-Methoxybenzyl)sulphamoyl]benzeneboronic acid 96%
3-[(4-Methoxybenzyl)sulphamoyl]benzeneboronic acid
Formel: C₁₄H₁₆BNO₅S
Molecular Weight: 321,157 g/mol
Schmelzpunkt: 110…112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08689541
CAS-Nummer: 913836-05-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR