5-(Methylthio)pyridine-3-boronic acid

Lieferant: Apollo Scientific
Achtung

OR3596-250MG
APOSOR3596-250MGEA 98.6 EUR
APOSOR3596-250MG
5-(Methylthio)pyridine-3-boronic acid
5-(Methylthio)pyridine-3-boronic acid
Formel: C₆H₈BNO₂S
Molecular Weight: 169,01 g/mol
Schmelzpunkt: 210…213 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08689538
CAS-Nummer: 477251-98-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR