Furan-2-boronsäurepinakolester 98%

Lieferant: Apollo Scientific

OR360000-5G
APOSOR360000-5GEA 37.9 EUR
APOSOR360000-5G
Furan-2-boronsäurepinakolester 98%
Furan-2-boronsäurepinakolester
Formel: C₁₀H₁₅BO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD02094008
CAS-Nummer: 374790-93-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR