3-(Morpholino)phenylboronsäurepinakolester

Lieferant: Apollo Scientific

OR360004-1G
APOSOR360004-1GEA 0 EUR
APOSOR360004-1G
3-(Morpholino)phenylboronsäurepinakolester
3-(Morpholino)phenylboronsäurepinakolester
Formel: C₁₆H₂₄BNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03412097
CAS-Nummer: 852227-95-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR