2-Methoxy-3-pyridinboronsäurepinakolester 97%
Lieferant: Apollo Scientific
Synonyme:
2-Methoxypyridine-3-boronic acid pinacol ester, 2-Methoxy-3-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyridine, 2-Methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
OR360013-5G
APOSOR360013-5GEA
102
EUR
APOSOR360013-5G
2-Methoxy-3-pyridinboronsäurepinakolester 97%
2-Methoxy-3-pyridinboronsäurepinakolester
Formel:
C₁₂H₁₈BNO₃ Molecular Weight: 235,09 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05663507 CAS-Nummer: 532391-31-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...