3-(p-Toluenesulphonylamino)benzeneboronic acid pinacol ester 97%
Lieferant: Apollo Scientific
OR360014-5G
APOSOR360014-5GEA
55.1
EUR
APOSOR360014-5G
3-(p-Toluenesulphonylamino)benzeneboronic acid pinacol ester 97%
3-(p-Toluenesulphonylamino)benzeneboronic acid pinacol ester
Formel:
C₁₉H₂₄BNO₄S Lagertemperatur: Raumtemperatur |
MDL:
MFCD05663863 CAS-Nummer: 796061-08-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...