3,5-Dichlorphenylboronsäurepinakolester 97%
Lieferant: Apollo Scientific
Synonyme:
2-(3,5-Dichlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
OR360018-25G
APOSOR360018-25GEA
150
EUR
APOSOR360018-25G
3,5-Dichlorphenylboronsäurepinakolester 97%
3,5-Dichlorphenylboronsäurepinakolester
Formel:
C₁₂H₁₅BCl₂O₂ Molecular Weight: 272.97 g/mol Schmelzpunkt: 53 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05863906 CAS-Nummer: 68716-51-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...