Benzocyclobutene-4-boronic acid

Lieferant: Apollo Scientific

Synonyme: Bicyclo[4.2.0]octa-1,3,5-trien-3-ylboronic acid

OR360030-1G OR360030-250MG
APOSOR360030-1GEA 348 EUR
APOSOR360030-1G APOSOR360030-250MG
Benzocyclobutene-4-boronic acid
Benzocyclobutene-4-boronic acid
Formel: C₈H₇BO₂
Molecular Weight: 145.951 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09752664
CAS-Nummer: 195730-31-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR