3-Propoxyphenylboronsäure 98%

Lieferant: Apollo Scientific

OR360035-5G
APOSOR360035-5GEA 66.7 EUR
APOSOR360035-5G
3-Propoxyphenylboronsäure 98%
3-Propoxyphenylboronsäure
Formel: C₉H₁₃BO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03427190
CAS-Nummer: 149557-18-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR