(4-Pyrrolidin-1-ylphenyl)boronic acid 95+%

Lieferant: Apollo Scientific

Synonyme: (4-(Pyrrolidin-1-yl)phenyl)boronic acid

OR360044-250MG
APOSOR360044-250MGEA 48 EUR
APOSOR360044-250MG
(4-Pyrrolidin-1-ylphenyl)boronic acid 95+%
(4-Pyrrolidin-1-ylphenyl)boronic acid
Formel: C₁₀H₁₄BNO₂
Molecular Weight: 191,035 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03095137
CAS-Nummer: 229009-41-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR