5-Cyano-2-methoxyphenylboronic acid 95+%

Lieferant: Apollo Scientific

Synonyme: (5-Cyano-2-methoxyphenyl)boronic acid

OR360058-1G
APOSOR360058-1GEA 112 EUR
APOSOR360058-1G
5-Cyano-2-methoxyphenylboronic acid 95+%
5-Cyano-2-methoxyphenylboronic acid
Formel: C₈H₈BNO₃
Molecular Weight: 176,97 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06801691
CAS-Nummer: 612833-37-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR