4-(1-Ethoxycarbonylcyclopropyl)phenylboronic acid, pinacol ester 95+%

Lieferant: Apollo Scientific

Synonyme: Ethyl 1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclopropanecarboxylate

OR360083-250MG
APOSOR360083-250MGEA 98.6 EUR
APOSOR360083-250MG
4-(1-Ethoxycarbonylcyclopropyl)phenylboronic acid, pinacol ester 95+%
4-(1-Ethoxycarbonylcyclopropyl)phenylboronic acid, pinacol ester
Formel: C₁₈H₂₅BO₄
Molecular Weight: 316.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18383419
CAS-Nummer: 1257213-52-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR