1-N-BOC-3-methyl-indazole-5-boronic acid, pinacol ester 97%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole-1-carboxylate

OR360108-250MG
APOSOR360108-250MGEA 56.7 EUR
APOSOR360108-250MG
1-N-BOC-3-methyl-indazole-5-boronic acid, pinacol ester 97%
1-N-BOC-3-methyl-indazole-5-boronic acid, pinacol ester
Formel: C₁₉H₂₇BN₂O₄
Molecular Weight: 358,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13195073
CAS-Nummer: 864770-82-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR