1-Cyclopentenylboronsäurepinakolester

Lieferant: Apollo Scientific
Achtung

OR360110-5G
APOSOR360110-5GEA 74.1 EUR
APOSOR360110-5G
1-Cyclopentenylboronsäurepinakolester
1-Cyclopentenylboronsäurepinakolester
Formel: C₁₁H₁₉BO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08669391
CAS-Nummer: 287944-10-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR