1-Ethylpyrazole-5-boronic acid, pinacol ester 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Ethylpyrazole-5-boronic Acid Pinacol Ester, 1-Ethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

OR360111-5G
APOSOR360111-5GEA 84.1 EUR
APOSOR360111-5G
1-Ethylpyrazole-5-boronic acid, pinacol ester 98%
1-Ethylpyrazole-5-boronic acid, pinacol ester
Formel: C₁₁H₁₉BN₂O₂
Molecular Weight: 222,092 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16659009
CAS-Nummer: 1007110-53-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR