Oxindol-6-boronsäurepinakolester 95+%

Lieferant: Apollo Scientific

Synonyme: 6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)indolin-2-one

OR360115-1G
APOSOR360115-1GEA 128 EUR
APOSOR360115-1G
Oxindol-6-boronsäurepinakolester 95+%
Oxindol-6-boronsäurepinakolester
Formel: C₁₄H₁₈BNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11855965
CAS-Nummer: 893441-85-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR