1-Phenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 97%

Lieferant: Apollo Scientific

Synonyme: 1-Phenylpyrazole-4-boronic acid, pinacol ester, 1-Phenyl-1H-pyrazole-4-boronic acid pinacol ester

OR360116-1G
APOSOR360116-1GEA 104 EUR
APOSOR360116-1G
1-Phenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 97%
1-Phenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Formel: C₁₅H₁₉BN₂O₂
Molecular Weight: 270,1346 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12400941
CAS-Nummer: 1002334-12-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR