3-((Dimethylamino)methyl)phenylboronic acid 95%
Lieferant: Apollo Scientific
Synonyme:
3-(N,N-Dimethylaminomethyl)phenylboronic acid
OR360120-1G
APOSOR360120-1GEA
106
EUR
APOSOR360120-1G
3-((Dimethylamino)methyl)phenylboronic acid 95%
3-((Dimethylamino)methyl)phenylboronic acid
Formel:
C₉H₁₄BNO₂ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
819849-22-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...