n-Pentylboronic acid 98%

Lieferant: Apollo Scientific

OR360127-25G
APOSOR360127-25GEA 79.9 EUR
APOSOR360127-25G
n-Pentylboronic acid 98%
n-Pentylboronic acid
Formel: C₅H₁₃BO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01074651
CAS-Nummer: 4737-50-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR