3-Ethylphenylboronic acid 97%
Lieferant: Apollo Scientific
OR360133-1G
APOSOR360133-1GEA
104
EUR
APOSOR360133-1G
3-Ethylphenylboronic acid 97%
3-Ethylphenylboronic acid
Formel:
C₈H₁₁BO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06657721 CAS-Nummer: 90555-65-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...