6-Hydroxy-3-pyridineboronic acid
Lieferant: Apollo Scientific
|
Gefahr
|
OR360140-1G
OR360140-250MG
APOSOR360140-1GEA
0
EUR
APOSOR360140-1G
APOSOR360140-250MG
6-Hydroxy-3-pyridineboronic acid
6-Hydroxy-3-pyridineboronic acid
Formel:
C₅H₆BNO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09907979 CAS-Nummer: 903899-13-8 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...