1-Cyclopropylmethyl-1H-pyrazole-4-boronic acid, pinacol ester 97%

Lieferant: Apollo Scientific

OR360142-1G
APOSOR360142-1GEA 127 EUR
APOSOR360142-1G
1-Cyclopropylmethyl-1H-pyrazole-4-boronic acid, pinacol ester 97%
1-Cyclopropylmethyl-1H-pyrazole-4-boronic acid, pinacol ester
Formel: C₁₃H₂₁BN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD15474887
CAS-Nummer: 1000801-75-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR