4-(2-Naphthyl)phenylboronsäure 95+%
Lieferant: Apollo Scientific
Synonyme:
(4-(Naphthalen-2-yl)phenyl)boronic acid
OR360155-25G
APOSOR360155-25GEA
194
EUR
APOSOR360155-25G
4-(2-Naphthyl)phenylboronsäure 95+%
4-(2-Naphthyl)phenylboronsäure
Formel:
C₁₆H₁₃BO₂ Molecular Weight: 248.0800 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09260454 CAS-Nummer: 918655-03-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...