2-Chlor-5-pyrimidinboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloropyrimidine-5-boronic acid

OR360157-25G
APOSOR360157-25GEA 59.6 EUR
APOSOR360157-25G
2-Chlor-5-pyrimidinboronsäure
2-Chloro-5-pyrimidineboronic acid
Formel: C₄H₄BClN₂O₂
Molecular Weight: 158.35 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08063113
CAS-Nummer: 1003845-06-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR