5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazolo[3,4-b]pyridine 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1H-Pyrazolo[3,4-b]pyridine-5-boronic acid, pinacol ester, 1H-Pyrazolo[3,4-b]pyridine-5-boronic acid pinacol ester
OR360160-250MG
APOSOR360160-250MGEA
55.1
EUR
APOSOR360160-250MG
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazolo[3,4-b]pyridine 95+%
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazolo[3,4-b]pyridine
Formel:
C₁₂H₁₆BN₃O₂ Molecular Weight: 245,0853 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09832892 CAS-Nummer: 1093819-50-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...