3-(tert-Butoxycarbonyl)phenylboronsäurepinakolester 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(t-Butoxycarbonyl)phenylboronic acid pinacol ester, tert-Butyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

OR360166-1G
APOSOR360166-1GEA 21.9 EUR
APOSOR360166-1G
3-(tert-Butoxycarbonyl)phenylboronsäurepinakolester 97%
3-(tert-Butoxycarbonyl)phenylboronsäurepinakolester
Formel: C₁₇H₂₅BO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD03453057
CAS-Nummer: 903895-48-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR