2-Methoxy-1-naphthalinboronsäure 98+%

Lieferant: Apollo Scientific

OR360184-1G
APOSOR360184-1GEA 65.4 EUR
APOSOR360184-1G
2-Methoxy-1-naphthalinboronsäure 98+%
2-Methoxy-1-naphthaleneboronic acid
Formel: C₁₁H₁₁BO₃
Molecular Weight: 202,0100 g/mol
Schmelzpunkt: 125…130 °C
Lagertemperatur: Kühlschrank
MDL: MFCD11044173
CAS-Nummer: 104116-17-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR