5-Methyl-4-bromothiophen-2-ylboronic acid 95+%

Lieferant: Apollo Scientific

Synonyme: 4-Bromo-5-methylthiophene-2-boronic acid, (4-Bromo-5-methylthiophen-2-yl)boronic acid

OR360220-250MG
APOSOR360220-250MGEA 150 EUR
APOSOR360220-250MG
5-Methyl-4-bromothiophen-2-ylboronic acid 95+%
5-Methyl-4-bromothiophen-2-ylboronic acid
Formel: C₅H₆BBrO₂S
Molecular Weight: 220,88 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09952079
CAS-Nummer: 154566-69-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR