2,4-Dimethoxyphenylboronsäurepinakolester 97%

Lieferant: Apollo Scientific

Synonyme: 2-(2,4-Dimethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR360230-5G
APOSOR360230-5GEA 84.1 EUR
APOSOR360230-5G
2,4-Dimethoxyphenylboronsäurepinakolester 97%
2,4-Dimethoxyphenylboronsäurepinakolester
Formel: C₁₄H₂₁BO₄
Molecular Weight: 264.13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06795688
CAS-Nummer: 214360-69-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR