2-Hydroxy-5-methylphenylboronic acid 97%

Lieferant: Apollo Scientific

Synonyme: (2-Hydroxy-5-methylphenyl)boronic acid

OR360235-1G
APOSOR360235-1GEA 114 EUR
APOSOR360235-1G
2-Hydroxy-5-methylphenylboronic acid 97%
2-Hydroxy-5-methylphenylboronic acid
Formel: C₇H₉BO₃
Molecular Weight: 151.956 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09839105
CAS-Nummer: 259209-21-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR