4-(Morpholinomethyl)phenylboronic acid 98%

Lieferant: Apollo Scientific

Synonyme: 4-(Morpholinomethyl)benzeneboronic acid, [4-(Morpholin-4-ylmethyl)phenyl]boronic acid

OR360236-1G
APOSOR360236-1GEA 62.5 EUR
APOSOR360236-1G
4-(Morpholinomethyl)phenylboronic acid 98%
4-(Morpholinomethyl)phenylboronic acid
Formel: C₁₁H₁₆BNO₃
Molecular Weight: 221,06 g/mol
Schmelzpunkt: 81…85 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06801707
CAS-Nummer: 279262-23-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR