3-Butoxyphenylboronic acid 97%

Lieferant: Apollo Scientific

OR360240-25G
APOSOR360240-25GEA 119 EUR
APOSOR360240-25G
3-Butoxyphenylboronic acid 97%
3-Butoxyphenylboronic acid
Formel: C₁₀H₁₅BO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03427053
CAS-Nummer: 352534-81-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR