3-Pyrrolidinophenylboronic acid 97%

Lieferant: Apollo Scientific

Synonyme: 3-(Pyrrolidino)phenylboronic acid

OR360259-250MG
APOSOR360259-250MGEA 79.9 EUR
APOSOR360259-250MG
3-Pyrrolidinophenylboronic acid 97%
3-Pyrrolidinophenylboronic acid
Formel: C₁₀H₁₄BNO₂
Molecular Weight: 191.035 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03095125
CAS-Nummer: 659731-18-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR