4-Cyano-3-methoxyphenylboronic acid 97%

Lieferant: Apollo Scientific

Synonyme: (4-Cyano-3-methoxyphenyl)boronic acid

OR360261-250MG
APOSOR360261-250MGEA 85.7 EUR
APOSOR360261-250MG
4-Cyano-3-methoxyphenylboronic acid 97%
4-Cyano-3-methoxyphenylboronic acid
Formel: C₈H₈BNO₃
Molecular Weight: 176,965 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10696655
CAS-Nummer: 677777-45-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR