4,4-Dimethylcyclohexen-1-ylboronsäurepinakolester

Lieferant: Apollo Scientific
Achtung

OR360276-1G OR360276-5G
APOSOR360276-1GEA 55.1 EUR
APOSOR360276-1G APOSOR360276-5G
4,4-Dimethylcyclohexen-1-ylboronsäurepinakolester
4,4-Dimethylcyclohexen-1-ylboronsäurepinakolester
Formel: C₁₄H₂₅BO₂
Lagertemperatur: Gefrierschrank
MDL: MFCD08690233
CAS-Nummer: 859217-67-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR